3-Bromo-5-fluoro-2-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

25960-1G 25960-25G 25960-5G
76912-126EA 94.43 USD
76912-126 76912-128 76912-130
3-Bromo-5-fluoro-2-iodotoluene ≥97%
3-Bromo-5-fluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Refrigerator
MDL Number: MFCD09800772
CAS Number: 1000576-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR