3,5-Dichloro-4-hydroxybenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
25926-25G
25926-5G
76912-114EA
300.24
USD
76912-114
76912-116
3,5-Dichloro-4-hydroxybenzaldehyde ≥97%
3,5-Dichloro-4-hydroxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00017605 CAS Number: 2314-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...