3-(N,N-Dimethylamino)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(N,N-Dimethylamino)phenylboronic acid, pinacol ester, N,N-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-(Dimethylamino)phenylboronic acid pinacol ester

25873-1G 25873-25G 25873-500MG 25873-5G
76912-088EA 162.32 USD
76912-088 76912-090 76912-092 76912-094
3-(N,N-Dimethylamino)phenylboronic acid pinacol ester ≥97%
3-(N,N-Dimethylamino)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₂
MW: 247.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795669
CAS Number: 325142-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR