3-(t-butyl)-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-(tert-Butyl)-4-methoxyphenyl)boronic acid

25832-1G 25832-25G 25832-500MG 25832-5G
76912-078EA 213.25 USD
76912-078 76912-080 76912-082 76912-084
3-(t-butyl)-4-methoxyphenylboronic acid ≥97%
3-(t-butyl)-4-methoxyphenylboronic acid
Formula: C₁₁H₁₇BO₃
MW: 208.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414597
CAS Number: 196960-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR