3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

25820-25G 25820-5G
76912-070EA 187.78 USD
76912-070 76912-072
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid ≥97%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃ClF₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01631509
CAS Number: 129931-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR