3-Bromo-2,4-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25655-1G 25655-25G 25655-500MG 25655-5G
76912-028EA 327.82 USD
76912-028 76912-030 76912-032 76912-034
3-Bromo-2,4-difluorobenzyl alcohol ≥97%
3-Bromo-2,4-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD28737915
CAS Number: 1499465-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR