4,5-Difluoro-2-iodoaniline ≥97% research grade

Supplier: AOBChem USA

Warning

25609-1G 25609-25G 25609-5G
76912-014EA 99.73 USD
76912-014 76912-016 76912-018
4,5-Difluoro-2-iodoaniline ≥97% research grade
4,5-Difluoro-2-iodoaniline
Formula: C₆H₄F₂IN
Storage Temperature: Ambient
MDL Number: MFCD03095321
CAS Number: 847685-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR