3-Bromo-5-(methylthio)benzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

25556-1G 25556-25G 25556-5G
76912-004EA 84.88 USD
76912-004 76912-006 76912-008
3-Bromo-5-(methylthio)benzotrifluoride ≥97%
3-Bromo-5-(methylthio)benzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₆BrF₃S
Storage Temperature: Ambient
MDL Number: MFCD11504946
CAS Number: 1072944-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR