4-(2-Pyridyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Pyridin-2-yl)phenylboronic acid, (4-(Pyridin-2-yl)phenyl)boronic acid

25484-1G 25484-25G 25484-5G
76911-990EA 74.27 USD
76911-990 76911-992 76911-994
4-(2-Pyridyl)phenylboronic acid ≥97%
4-(2-Pyridyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₀BNO₂
MW: 199.014 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17013972
CAS Number: 170230-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR