Methyl 3-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-fluorobenzoic acid methyl ester

25455-25G 25455-5G
76911-982EA 488.02 USD
76911-982 76911-984
Methyl 3-chloro-5-fluorobenzoate ≥97%
Methyl 3-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD06204128
CAS Number: 327056-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR