3,5-Dibromo-2-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

25226-1G 25226-25G 25226-5G
76911-954EA 73.21 USD
76911-954 76911-956 76911-958
3,5-Dibromo-2-iodotoluene ≥97%
3,5-Dibromo-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂I
Storage Temperature: Refrigerator
MDL Number: MFCD09800828
CAS Number: 1000571-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR