4-(5-Bromo-2-fluorobenzyl)morpholine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-fluorophenyl-2-(morphlinomethyl)benzene

25191-1G 25191-25G 25191-5G
76911-928EA 73.21 USD
76911-928 76911-930 76911-932
4-(5-Bromo-2-fluorobenzyl)morpholine ≥95%
4-(5-Bromo-2-fluorobenzyl)morpholine
Formula: C₁₁H₁₃BrFNO
MW: 274.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441943
CAS Number: 488799-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR