3-Formyl-4-methylbenzonitrle ≥95%

Supplier: AOBChem USA

Warning

25110-1G 25110-25G 25110-500MG 25110-5G
76911-912EA 381.93 USD
76911-912 76911-914 76911-916 76911-918
3-Formyl-4-methylbenzonitrle ≥95%
3-Formyl-4-methylbenzonitrle
Formula: C₉H₇NO
MW: 145.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18394299
CAS Number: 27613-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR