3-Bromo-4-(hydroxymethyl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

25018-1G 25018-25G 25018-5G
76911-894EA 117.76 USD
76911-894 76911-896 76911-898
3-Bromo-4-(hydroxymethyl)benzonitrile ≥97%
3-Bromo-4-(hydroxymethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD09907613
CAS Number: 90110-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR