3-Fluoro-5-phenylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

24891-1G 24891-25G 24891-500MG 24891-5G
76911-858EA 290.69 USD
76911-858 76911-860 76911-862 76911-864
3-Fluoro-5-phenylpyridine ≥97% research grade
3-Fluoro-5-phenylpyridine
Formula: C₁₁H₈FN
Storage Temperature: Refrigerator
MDL Number: MFCD14701970
CAS Number: 1214374-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR