(3-Bromo-4-chlorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

24856-1G 24856-25G 24856-500MG 24856-5G
76911-832EA 144.29 USD
76911-832 76911-834 76911-836 76911-838
(3-Bromo-4-chlorophenyl)boronic acid ≥95%
(3-Bromo-4-chlorophenyl)boronic acid
Formula: C₆H₅BBrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD19981540
CAS Number: 1384956-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR