3-Bromo-5-[(pyrrolidin-1-yl)methyl]pyridine ≥97%

Supplier: AOBChem USA

Warning

24738-1G 24738-25G 24738-500MG 24738-5G
76911-804EA 290.69 USD
76911-804 76911-806 76911-808 76911-810
3-Bromo-5-[(pyrrolidin-1-yl)methyl]pyridine ≥97%
3-Bromo-5-[(pyrrolidin-1-yl)methyl]pyridine
Formula: C₁₀H₁₃BrN₂
MW: 241.128 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22628211
CAS Number: 183608-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR