3-Bromo-5-chlorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-chlorophenyl)methanol

24724-25G 24724-5G
76911-794EA 502.87 USD
76911-794 76911-796
3-Bromo-5-chlorobenzyl alcohol ≥95%
3-Bromo-5-chlorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD08061830
CAS Number: 917562-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR