3-Ethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24717-25G 24717-5G
76911-784EA 360.71 USD
76911-784 76911-786
3-Ethylbenzaldehyde ≥97%
3-Ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09263482
CAS Number: 34246-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR