3-Cyclopropyl-2-fluoropyridine ≥97%

Supplier: AOBChem USA

Warning

24685-1G 24685-25G 24685-500MG 24685-5G
76911-762EA 127.31 USD
76911-762 76911-764 76911-766 76911-768
3-Cyclopropyl-2-fluoropyridine ≥97%
3-Cyclopropyl-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₈H₈FN
Storage Temperature: Refrigerator
MDL Number: MFCD20528324
CAS Number: 1227177-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR