3-Bromo-5-Chloro-2-(Methylsulfanyl)Pyridine ≥97%

Supplier: AOBChem USA

Warning

24648-25G 24648-5G
76911-754EA 346.92 USD
76911-754 76911-756
3-Bromo-5-Chloro-2-(Methylsulfanyl)Pyridine ≥97%
3-Bromo-5-Chloro-2-(Methylsulfanyl)Pyridine
Formula: C₆H₅BrClNS
MW: 238.533 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543737
CAS Number: 1289214-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR