3-Bromo-5-chloro-thioanisole ≥97%

Supplier: AOBChem USA

Warning

24562-25G 24562-5G
76911-686EA 300.24 USD
76911-686 76911-688
3-Bromo-5-chloro-thioanisole ≥97%
3-Bromo-5-chloro-thioanisole
Formula: C₇H₆BrClS
MW: 237.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488914
CAS Number: 1394291-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR