4-Bromo-2-chloro-6-methylphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24532-1G 24532-500MG 24532-5G
76911-648EA 668.37 USD
76911-648 76911-650 76911-652
4-Bromo-2-chloro-6-methylphenylboronic acid ≥97% research grade
4-Bromo-2-chloro-6-methylphenylboronic acid
Formula: C₇H₇BBrClO₂
Storage Temperature: Ambient
MDL Number: MFCD22543721
CAS Number: 1451391-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR