3-Bromo-2-methyl-6-(2-methylpropoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

24451-1G 24451-25G 24451-500MG 24451-5G
76911-604EA 162.32 USD
76911-604 76911-606 76911-608 76911-610
3-Bromo-2-methyl-6-(2-methylpropoxy)pyridine ≥97%
3-Bromo-2-methyl-6-(2-methylpropoxy)pyridine
Formula: C₁₀H₁₄BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD22628193
CAS Number: 1289124-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR