3,5-Difluoro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24335-1G 24335-25G 24335-500MG 24335-5G
76911-544EA 142.17 USD
76911-544 76911-546 76911-548 76911-550
3,5-Difluoro-4-iodobenzaldehyde ≥97%
3,5-Difluoro-4-iodobenzaldehyde
Formula: C₇H₃F₂IO
Storage Temperature: Refrigerator
MDL Number: MFCD22543693
CAS Number: 357166-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR