3,5-Dibromo-4-methoxypyridine ≥97% research grade

Supplier: AOBChem USA

Warning

24311-25G 24311-5G
76911-532EA 219.61 USD
76911-532 76911-534
3,5-Dibromo-4-methoxypyridine ≥97% research grade
3,5-Dibromo-4-methoxypyridine
Formula: C6H5Br2NO
Storage Temperature: Ambient
MDL Number: MFCD13185524
CAS Number: 25813-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR