2,6-Dichloro-4-(trifluoromethyl)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

24258-1G 24258-25G 24258-500MG 24258-5G
76911-522EA 171.87 USD
76911-522 76911-524 76911-526 76911-528
2,6-Dichloro-4-(trifluoromethyl)benzoic acid ≥97% research grade
2,6-Dichloro-4-(trifluoromethyl)benzoic acid
Formula: C₈H₃Cl₂F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD01934568
CAS Number: 189338-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR