3-Bromo-2,4-difluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
24220-1G
24220-25G
24220-5G
76911-510EA
94.43
USD
76911-510
76911-512
76911-514
3-Bromo-2,4-difluorobenzoic acid ≥97%
3-Bromo-2,4-difluorobenzoic acid
Formula:
C₇H₃BrF₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09261257 CAS Number: 651026-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...