3-Chloro-2-(Methylsulfanyl)Phenol ≥97%

Supplier: AOBChem USA

Warning

24192-1G 24192-25G 24192-500MG 24192-5G
76911-496EA 194.15 USD
76911-496 76911-498 76911-500 76911-502
3-Chloro-2-(Methylsulfanyl)Phenol ≥97%
3-Chloro-2-(Methylsulfanyl)Phenol
Formula: C₇H₇ClOS
MW: 174.648 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16998897
CAS Number: 406935-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR