(5-chloro-6-fluoropyridin-3-yl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-fluoropyridne-5-boronic acid, (5-Chloro-6-fluoropyridin-3-yl)boronic acid

24024-1G 24024-25G 24024-500MG 24024-5G
76911-458EA 165.51 USD
76911-458 76911-460 76911-462 76911-464
(5-chloro-6-fluoropyridin-3-yl)boronic acid ≥97% research grade
(5-chloro-6-fluoropyridin-3-yl)boronic acid
Formula: C₅H₄BClFNO₂
MW: 175.353 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257900
CAS Number: 1366482-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR