4'-Fluoro-biphenyl-3-carbaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
4'-Fluoro-[1,1'-biphenyl]-3-carbaldehyde, 3-(4-Fluorophenyl)benzaldehyde
23928-1G
23928-25G
23928-5G
76911-434EA
76.39
USD
76911-434
76911-436
76911-438
4'-Fluoro-biphenyl-3-carbaldehyde ≥95%
4'-Fluoro-biphenyl-3-carbaldehyde
Formula:
C₁₃H₉FO MW: 200.21 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01631878 CAS Number: 164334-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...