4-Benzyloxy-5-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23806-1G 23806-25G 23806-500MG 23806-5G
76911-382EA 142.17 USD
76911-382 76911-384 76911-386 76911-388
4-Benzyloxy-5-fluoro-3-methoxybenzaldehyde ≥97%
4-Benzyloxy-5-fluoro-3-methoxybenzaldehyde
Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28977121
CAS Number: 99387-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR