3-Bromo-5-fluoro-4-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

23803-1G 23803-25G 23803-5G
76911-368EA 64.72 USD
76911-368 76911-370 76911-372
3-Bromo-5-fluoro-4-iodotoluene ≥97%
3-Bromo-5-fluoro-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD09800758
CAS Number: 1000576-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR