3-Chloro-2-(Methylsulfanyl)Phenylboronic Acid ≥97%

Supplier: AOBChem USA

Warning

23742-1G 23742-25G 23742-500MG 23742-5G
76911-348EA 180.36 USD
76911-348 76911-350 76911-352 76911-354
3-Chloro-2-(Methylsulfanyl)Phenylboronic Acid ≥97%
3-Chloro-2-(Methylsulfanyl)Phenylboronic Acid
Formula: C₇H₈BClO₂S
Storage Temperature: Ambient
MDL Number: MFCD29088124
CAS Number: 2121513-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR