3-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine, 3-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester

23644-25G 23644-5G
76911-336EA 300.24 USD
76911-336 76911-338
3-(Morpholinomethyl)phenylboronic acid pinacol ester ≥97% research grade
3-(Morpholinomethyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD06797489
CAS Number: 364794-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR