3-Bromo-4-iodothiophene ≥95%

Supplier: AOBChem USA

Warning

23608-1G 23608-25G 23608-500MG 23608-5G
76911-312EA 120.95 USD
76911-312 76911-314 76911-316 76911-318
3-Bromo-4-iodothiophene ≥95%
3-Bromo-4-iodothiophene
Formula: C₄H₂BrIS
Storage Temperature: Refrigerator
MDL Number: MFCD18382466
CAS Number: 73882-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR