4,5-Difluoro-2-methoxyphenol ≥95% research grade

Supplier: AOBChem USA

Warning

23420-1G 23420-25G 23420-5G
76911-250EA 84.88 USD
76911-250 76911-252 76911-254
4,5-Difluoro-2-methoxyphenol ≥95% research grade
4,5-Difluoro-2-methoxyphenol
Formula: C₇H₆F₂O₂
MW: 160.118 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706859
CAS Number: 246029-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR