4,5-Difluoro-2-hydroxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23362-1G 23362-25G 23362-500MG 23362-5G
76911-234EA 173.99 USD
76911-234 76911-236 76911-238 76911-240
4,5-Difluoro-2-hydroxybenzoic acid ≥97%
4,5-Difluoro-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD13185554
CAS Number: 205533-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR