4,5-Difluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23358-25G 23358-5G
76911-222EA 350.1 USD
76911-222 76911-224
4,5-Difluoro-2-methoxybenzoic acid ≥97%
4,5-Difluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660196
CAS Number: 425702-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR