4-(Benzyloxy)-2-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23330-1G 23330-25G 23330-500MG 23330-5G
76911-210EA 260.99 USD
76911-210 76911-212 76911-214 76911-216
4-(Benzyloxy)-2-chlorobenzoic acid ≥97%
4-(Benzyloxy)-2-chlorobenzoic acid
Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02676704
CAS Number: 75835-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR