(4-(4-Fluorophenoxy)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

23328-1G 23328-25G 23328-5G
76911-198EA 88.06 USD
76911-198 76911-200 76911-202
(4-(4-Fluorophenoxy)phenyl)boronic acid ≥97%
(4-(4-Fluorophenoxy)phenyl)boronic acid
Formula: C₁₂H₁₀BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13182406
CAS Number: 361437-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR