4-Benzyloxy-2-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

23311-25G 23311-5G
76911-190EA 368.14 USD
76911-190 76911-192
4-Benzyloxy-2-chlorophenylboronic acid ≥97%
4-Benzyloxy-2-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
Storage Temperature: Ambient
MDL Number: MFCD06801734
CAS Number: 1315341-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR