2-Amino-6-iodotoluene ≥95%

Supplier: AOBChem USA

Warning

23189-25G 23189-5G
76911-172EA 375.56 USD
76911-172 76911-174
2-Amino-6-iodotoluene ≥95%
2-Amino-6-iodotoluene
Formula: C₇H₈IN
Storage Temperature: Ambient
MDL Number: MFCD08062502
CAS Number: 172681-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR