3-Bromo-5-cyclopropylpyridine ≥97%

Supplier: AOBChem USA

Warning

23121-1G 23121-25G 23121-500MG 23121-5G
76911-144EA 245.07 USD
76911-144 76911-146 76911-148 76911-150
3-Bromo-5-cyclopropylpyridine ≥97%
3-Bromo-5-cyclopropylpyridine
Formula: C₈H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD11870213
CAS Number: 1044210-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR