6-Fluoroveratraldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde

23040-25G 23040-5G
76911-110EA 222.79 USD
76911-110 76911-112
6-Fluoroveratraldehyde ≥97%
6-Fluoroveratraldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061108
CAS Number: 71924-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR