4,5-Dibromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22973-1G 22973-25G 22973-500MG 22973-5G
76911-096EA 439.22 USD
76911-096 76911-098 76911-100 76911-102
4,5-Dibromo-1H-indazole ≥97%
4,5-Dibromo-1H-indazole
Formula: C₇H₄Br₂N₂
MW: 275.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526398
CAS Number: 1351668-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR