3,5-Dibromo-4-(1,3-dioxolan-2-yl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

22964-25G 22964-5G
76911-084EA 519.85 USD
76911-084 76911-086
3,5-Dibromo-4-(1,3-dioxolan-2-yl)pyridine ≥97% research grade
3,5-Dibromo-4-(1,3-dioxolan-2-yl)pyridine
Formula: C₈H₇Br₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD25969059
CAS Number: 924649-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR