3-(t-Butyldimethylsilyloxy)phenylboronic acid, pinacol ester ≥95%

Supplier: AOBChem USA

Warning

22952-1G 22952-25G 22952-5G
76911-072EA 64.72 USD
76911-072 76911-074 76911-076
3-(t-Butyldimethylsilyloxy)phenylboronic acid, pinacol ester ≥95%
3-(t-Butyldimethylsilyloxy)phenylboronic acid, pinacol ester
Formula: C₁₈H₃₁BO₃Si
MW: 334.333 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04039307
CAS Number: 902120-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR