2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-cyanophenylboronic acid neopentyl glycol ester

22883-1G 22883-25G 22883-5G
76911-058EA 91.24 USD
76911-058 76911-060 76911-062
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile ≥95%
2-Chloro-6-(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzonitrile
Formula: C₁₂H₁₃BClNO₂
MW: 249.50 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669560
CAS Number: 883899-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR