4-(3-Bromo-4-fluorobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(3-Bromo-5-fluorobenzyl)morpholine

22850-1G 22850-25G 22850-500MG 22850-5G
76911-042EA 159.14 USD
76911-042 76911-044 76911-046 76911-048
4-(3-Bromo-4-fluorobenzyl)morpholine ≥97%
4-(3-Bromo-4-fluorobenzyl)morpholine
Formula: C₁₁H₁₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD01307672
CAS Number: 281652-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR